Order Entry
Spain
ContactUsLinkComponent
VWR®, Ring Lights for VisiLight® LED
VWR®, Ring Lights for VisiLight® LED
Catalog # 631-1753
Supplier:  VWR Collection
undefined
VWR®, Ring Lights for VisiLight® LED
Catalog # 631-1753
Supplier:  VWR Collection

Specifications

  • Description:
    Fibre optic ring light (inner Ø = 66 mm / L = 750 mm)
  • Seq:
    500
  • Pk:
    1

Specifications

About this item