Anti-IgM Antibody (HRP (Horseradish Peroxidase))
Catalog # BSBTBA1075-0.25
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Conjugation:HRP (Horseradish Peroxidase)
- Cross adsorption:No
- Size:250 μg
- Pk:0,25 mg
Specifications
About this item
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: