Anti- IgG Antibody (Unconjugated)
Catalog # BSBTBA1038-5
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Unconjugated
- Cross adsorption:No
- Size:5 mg
- Pk:5 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: