Order Entry
Spain
ContactUsLinkComponent
 
Anti-Protein A Antibody (Biotin)
  BSBTBA1025-0.5
Anti-Protein A Antibody (Biotin)
  BSBTBA1025-0.5

 

  • Antibody type:
    Tag
  • Antigen name:
    Protein A
  • Conjugation:
    Biotin
  • Cross adsorption:
    No
  • Size:
    500 μg
  • Pk:
    0,5 mg

 

 

Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: