Anti-NPHS2 Rabbit Antibody
BSBTPA1322-1
- Antibody type:Primary
- Antigen name:Nephrosis 2, idiopathic, steroid-resistant (podocin)
- Antigen symbol:NPHS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:368 to 383
- Sequence:KPVEPLNPKKKDSPML
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human NPHS2 (368-383aa KPVEPLNPKKKDSPML), identical to the related mouse sequence, and different from the related rat sequence by one amino acid.
- Purification:Purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Podocin(NPHS2) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: NPHS2
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat