Order Entry
Spain
ContactUsLinkComponent
Grease, vacuum, Apiezon®
Grease, vacuum, Apiezon®
Catalog # EDWAH11150001
Grease, vacuum, Apiezon®
Catalog # EDWAH11150001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Apiezon® greases
  • Pk:
    25 g
Specifications
About this item
A range of Apiezon® greases is offered for use on high vacuum systems and for many general applications in the laboratory. These high purity greases are non-toxic, are easy to apply and clean off and have long shelf lives.
  • With good glittering qualities
  • Non-vacuum uses include lubrication of glass and metal taps in laboratory, medical and industrial applications