Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Entrada de pedido
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Medios
Medios preparados
Culture media, ready to use, in plates
Culture media, ready to use, in plates
Culture media, ready to use, in plates
Catalog # DIFC257635
Supplier:
DIFCO BD
CAS Number:
Expand image
Culture media, ready to use, in plates
Catalog # DIFC257635
Supplier:
DIFCO BD
CAS Number:
Specifications
Env:
100
Estándar:
-
Descripción:
Trypticase Soy agar+Lecithin+Polysorb 80
Specifications
About this item
Product Details
Documentation
Product Details
Documentation