BD BBL™ Prepared tubed and Mycoflask media
Supplier: DIFCO BD
Certificates
About this item
BBL™ Prepared Tubed Media are for the isolation and identification of microorganisms. Tubed media are ideal for cultures requiring prolonged incubation, since many can be incubated with caps tightly closed to prevent dehydration and subsequent inhibition of growth.
- Reduce possible contamination
- Special long-necked, rubber lined screw caps eliminate danger of contamination
- Larger-sized tubes with wider diameters permit easy inoculation and handling of media
Frequently Bought Together