Order Entry
Spain
Orders LinkContactUsLinkComponent
Papain

Papain

25 kg
Catalog # 1.07147.9025
Papain
Catalog # 1.07147.9025
CAS Number:  
Papain

25 kg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    25 kg
  • Cat. No.:
    1.07147.9025
  • Enzyme activity:
    6000 USP-U/mg
  • Tested applications:
    Suitable for use as excipient
  • Enzyme name:
    Papain

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.