
Vitamin A water miscible, 1 ml = 200,000 I.U stabilised, water-miscible, EMPROVE® ESSENTIAL BP, Ph. Eur., SAFC®
:68-26-8
:233-466-0
:GHS08
:Ambient
- Appearance:clear, yellow, viscous liquid
- Identity:passes test
- Miscibility with water:passes test
- pH-value (10 %; water):6.5 - 7.5
- Assay on vitamin A-palmitate:190000 - 230000 I.U./ml
- Density (d 20 °C/ 4 °C):ca. 1.04
- Microbiological test:passes test
- Residual solvents (Ph. Eur./ICH):excluded by production process
Frequently Bought Together