Anti-ZO-2, Mid Rabbit Polyclonal Antibody
Catalog # USBIZ0760-01B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:ZO-2, Mid
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-3?g/ml; Immunofluorescence: 2.5 ?g/ml
Type: Primary
Antigen: ZO-2, Mid
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human