Anti-PRPF4B Rabbit Polyclonal Antibody
Catalog # USBIP9120-84
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:pre-mRNA processing factor 4B
- Antigen symbol:PRPF4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: PRPF4B (pre-mRNA processing factor 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human