Anti-Platelet Derived Growth Factor BB Rabbit Polyclonal Antibody
Catalog # USBIP4258-17F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Platelet Derived Growth Factor BB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,1M Tris-HCl, pH 7,5.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Platelet Derived Growth Factor BB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human