Anti-NIP7 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIN2567-87C-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NIP7, nucleolar pre-rRNA processing Protein
- Antigen symbol:NIP7
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: NIP7 (NIP7, nucleolar pre-rRNA processing protein)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human