Anti-Muscarinic Acetylcholine Receptor M3 Rabbit Polyclonal Antibody
USBIM9520-23D1
- Antibody type:Primary
- Antigen name:Cholinergic receptor, muscarinic 3
- Antigen symbol:CHRM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Muscarinic Acetylcholine Receptor M3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human