Anti-Junctional Adhesion Molecule 4 Sheep Polyclonal Antibody (Biotin)
USBIJ8009-30
- Antibody type:Primary
- Antigen name:Junctional Adhesion Molecule 4
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Sheep
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:16 to 140
- Storage buffer:PBS containing 50ug of BSA per 1ug of antibody.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: Junctional Adhesion Molecule 4
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse