Anti-Integrin beta-like 1 with EGF-like Repeat Domains Chicken Polyclonal Antibody
Catalog # USBII7661-49
Proveedor: US Biological
Especificaciones
- Antibody type:Primary
- Antigen name:Integrin beta-like 1 with EGF-like Repeat Domains
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- ImmunoChemistry:Yes
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:242 to 291
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Especificaciones
Acerca de este artículo
Recommended dilution: Western Blot: 1:250; Immunochemistry: 1:100;
Type: Primary
Antigen: Integrin beta-like 1 with EGF-like Repeat Domains
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human