Anti-INCENP Rabbit Polyclonal Antibody
Catalog # USBII0795-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:inner centromere Protein antigens 135/155 kDa
- Antigen symbol:INCENP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:10 mM HEPES (pH 7,5), 150 mM NaCl, 100ug/mlBSA and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by peptide affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:400; Flow Cytometry: 1:50; Western Blot: 1:1000
Type: Primary
Antigen: INCENP (inner centromere protein antigens 135/155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human