Anti-Heat Shock Protein 22 Goat Polyclonal Antibody
Catalog # USBIH1830-48M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein 22
- Antigen symbol:HSP22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:184 to 196
- Storage buffer:TBS, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Heat Shock Protein 22
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human