Anti-AGO1 Rabbit Polyclonal Antibody
Catalog # USBIE2237-25B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:argonaute RISC catalytic component 1
- Antigen symbol:AGO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: AGO1 (argonaute RISC catalytic component 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat