Anti-ITGAE (Integrin alpha E, CD103) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 3H1797]
USBIC2445-63L
- Antibody type:Primary
- Antigen name:Integrin Alpha E
- Antigen symbol:ITGAE
- Clonality:Monoclonal
- Clone:3H1797
- Conjugation:PE (Phycoerythrin)/Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: ITGAE (Integrin alpha E, CD103)
Clonality: Monoclonal
Clone: 3H1797
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human