Anti-ICAM2 Mouse Monoclonal Antibody [clone: CBR-IC2/2]
Catalog # USBIC2445-44G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:intercellular adhesion molecule 2
- Antigen symbol:ICAM2
- Clonality:Monoclonal
- Clone:CBR-IC2/2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ICAM2 (intercellular adhesion molecule 2)
Clonality: Monoclonal
Clone: CBR-IC2/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human