Order Entry
Spain
ContactUsLinkComponent
Anti-SERPINF2 Mouse Monoclonal Antibody [clone: 5F134]
Anti-SERPINF2 Mouse Monoclonal Antibody [clone: 5F134]
Catalog # USBIA2298-13J
Supplier:  US Biological
Anti-SERPINF2 Mouse Monoclonal Antibody [clone: 5F134]
Catalog # USBIA2298-13J
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade F, member 2
  • Antigen symbol:
    SERPINF2
  • Clonality:
    Monoclonal
  • Clone:
    5F134
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,4, 20mg/ml mannitol, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation followed by fast polymer liquid chromatography (FPLC) separation
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: SERPINF2 (serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2)
Clonality: monoclonal
Clone: 5F134
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human