Order Entry
Spain
ContactUsLinkComponent
 
Anti-Amyloid, Oligomer A11 Rabbit Polyclonal Antibody
  USBIA2275-68A
undefined
Anti-Amyloid, Oligomer A11 Rabbit Polyclonal Antibody
  USBIA2275-68A

 

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid, Oligomer A11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:500; Immunoprecipitation: 1:1000

Type: Primary
Antigen: Amyloid, Oligomer A11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat