Anti-LEDGF Mouse Monoclonal Antibody [clone: 762815]
Catalog # USBI145504
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:LEDGF
- Clonality:Monoclonal
- Clone:762815
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:1 to 333
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: LEDGF
Clonality: monoclonal
Clone: 762815
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human