Anti-ABCC11 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI031475-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 11
- Antigen symbol:ABCC11
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:350 to 380
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: ABCC11 (ATP-binding cassette, sub-family C (CFTR/MRP), member 11)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human