Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Catalog # ROCK803-4902
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Texas Red®
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Chicken
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Chicken IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Size:500 µl
- Pk:500 µG
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Chicken