Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Catalog # ROCK803-4602
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Chicken
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Chicken IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Suitable for immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer specificity.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Chicken