Anti-IgG (H+L) Donkey Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ROCK709-708-123
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:0,5 mg/ml by absorbance = 82,0 at 565 nm
- Shipping temperature:Ice
- Immunogen:Human IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250;
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Human