Anti-IgG (H+L) Mouse Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
ROCK205-3202
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Goat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:20 mg
- Pk:20 mg
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Mouse
Isotype: IgG
Reactivity: Goat