Anti-CDK1 Rabbit Monoclonal Antibody (Unconjugated) [clone: 10H11.E12]
ROCK100-401-160
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:983
- Antigen synonyms:P34CDC2|CDC2|CDC28A
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:cdc2 peptide corresponding to the C-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
This antibody is suitable for immunoblotting, immunochemistry immuno-precipitation (as active kinase), and immunoblotting. HeLa cell lysate or human colon carcinoma is suggested as a positive control for immunoblotting.
Recommended Dilutions: ELISA: 1:60,000; Western Blot: 1:250 - 1:5000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CDK1
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human