Anti-Biotin Goat Monoclonal Antibody (Unconjugated) [clone: 9F9.F9]
ROCK100-1198
- Antibody type:Tag
- Antigen name:Biotin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.
Recommended Dilutions: ELISA: 1:100000 - 1:500,000; Western Blot: 1:10000 - 1:50000;
Restore with deionized water (or equivalent); 2.0 ml
Type: Tag
Antigen: Biotin
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan