Anti-Tissue Plasminogen Activator (Ab-1) (GMA-043) Mouse Monoclonal Antibody [clone: GMA-043]
GF53-100
- Antibody type:Primary
- Antigen name:Tissue Plasminogen Activator
- Antigen symbol:TPA
- Clonality:Monoclonal
- Clone:GMA-043
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Tissue Plasminogen Activator (Ab-1) (GMA-043)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human