Anti-Superoxide Dismutase (Cu/Zn Enzyme) Sheep Polyclonal Antibody
Catalog # 574597-1
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:superoxide dismutase Cu/Zn Enzyme
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Superoxide Dismutase (Cu/Zn Enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: