Specifications
- Antibody type:Primary
- Antigen name:Arrestin Beta 2
- Antigen symbol:ARB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:C-arrestin |cArr|ARRX|X-arrestin |CAR
- Amino acid number:395 to 409
- UniProtKB:P32121
- Sequence:RLKGMKDDDYDDQLC
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Beta Arrestin 2 (395-409aa RLKGMKDDDYDDQLC), different from the related rat and mouse sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Beta-arrestin-2(ARRB2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
ARRB2(arrestin beta-2), also known as Beta-arrestin-2, BARR2, ARB2, DKFZp686L0365, arrestin 3, is an intracellularprotein that in humans is encoded by the ARRB2 gene. Members of arrestin/beta-arrestin protein family are though to participate in agonist-mediated desensitization of G protein-coupled receptors and cause specific dampening of cellular responses to stimuli such as hormones, neurotransmitters, or sensory signals, as well as having signalling roles in their own right. Arrestin beta 2, like arrestin beta 1, was shown to inhibit beta-adrenergic receptor function in vitro. It is expressed at high levels in the central nervous system and may play a role in the regulation of synaptic receptors. Beta-arrestins were originally discovered in the context of heterotrimeric G protein-coupled receptor desensitization, but they also function in internalization and signaling of these receptors.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
WB: The detection limit for ARRB2 is approximately 5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Beta Arrestin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat