Anti-SPINK1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: SPINK1 (serine peptidase inhibitor, Kazal type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
- Catalog No:
- NOVUNBP2-29620
- NOVUNBP2-29620SS
- Antigen symbol:
- SPINK1
- SPINK1
- Antigen name:
- Serine Peptidase Inhibitor, Kazal Type 1
- Serine Peptidase Inhibitor, Kazal Type 1
- Antigen synonyms:
- Spink3|TATI|TCP|PCTT|PSTI
- Spink3|TATI|TCP|PCTT|PSTI
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6690
- 6690
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together