Order Entry
Spain
ContactUsLinkComponent
 
Anti-CD33 Rabbit Polyclonal Antibody
 
undefined
Anti-CD33 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNBP2-29619
  • NOVUNBP2-29619SS
  • Antigen symbol:
  • CD33
  • CD33
  • Antigen name:
  • CD33 molecule
  • CD33 molecule
  • Antigen synonyms:
  • SIGLEC-3|SIGLEC3|p67
  • SIGLEC-3|SIGLEC3|p67
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 945
  • 945
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together