Anti-MYD88 Mouse Monoclonal Antibody [clone: 4D6]
NOVUNBP2-27369
- Antibody type:Primary
- Antigen name:Myeloid differentiation primary response 88
- Antigen symbol:MYD88
- Clonality:Monoclonal
- Clone:4D6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:4615
- Antigen synonyms:MYD88D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: monoclonal
Clone: 4D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human