Anti-S1PR1 Rabbit Polyclonal Antibody
NOVUNBP2-27291
- Antibody type:Primary
- Antigen name:sphingosine-1-phosphate receptor 1
- Antigen symbol:S1PR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Gene ID:1901
- Antigen synonyms:CD363|CHEDG1|ECGF1|D1S3362|EDG-1|EDG1|S1P1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: S1PR1 (sphingosine-1-phosphate receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human