Anti-ARHGEF11 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24517
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Rho guanine nucleotide exchange factor (GEF) 11
- Antigen symbol:ARHGEF11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:9826
- Antigen synonyms:GTRAP48|PDZ-RHOGEF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ARHGEF11 (Rho guanine nucleotide exchange factor (GEF) 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: