Order Entry
Spain
ContactUsLinkComponent
Anti-RSAD2 Rabbit Polyclonal Antibody
Anti-RSAD2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-03971
Supplier:  Novus Biologicals
Anti-RSAD2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-03971
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Viperin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    91543
  • Antigen synonyms:
    vig1|cig5|2510004L01Rik|cig33
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: RSAD2 (radical S-adenosyl methionine domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: