Anti-SETX Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-94712C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Senataxin
- Antigen symbol:SETX
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:23064
- Antigen synonyms:bA479K20.2|ALS4|AOA2|SCAR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SETX (senataxin)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human