Anti-NLRC4 Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNBP1-78980G
- Antibody type:Primary
- Antigen name:NLR family, CARD domain containing 4
- Antigen symbol:NLRC4
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:58484
- Antigen synonyms:CLANA|AIFEC|CLAN|FCAS4|CARD12|CLAN1|CLR2.1|CLANC|IPAF|CLAND|CLANB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: NLRC4 (NLR family, CARD domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse