Anti-MAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
NOVUNBP1-78444H
- Antibody type:Primary
- Antigen name:MAS1 oncogene
- Antigen symbol:MAS1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:4142
- Antigen synonyms:MGC119966|MGRA|MAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: MAS1 (MAS1 oncogene)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: