Order Entry
Spain
ContactUsLinkComponent
Anti-FAT4 Rabbit Polyclonal Antibody
Anti-FAT4 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-FAT4 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-78381SS
  • NOVUNBP1-78381
  • Antigen symbol:
  • FAT4
  • FAT4
  • Antigen name:
  • FAT atypical cadherin 4
  • FAT atypical cadherin 4
  • Antigen synonyms:
  • CDHR11|CDHF14|HKLLS2|FATJ|VMLDS2|NBLA00548|FAT-J
  • CDHR11|CDHF14|HKLLS2|FATJ|VMLDS2|NBLA00548|FAT-J
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 79633
  • 79633
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together