Order Entry
Spain
ContactUsLinkComponent
 
Anti-KRT19 Rabbit Polyclonal Antibody
 
undefined
Anti-KRT19 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNBP1-78278
  • NOVUNBP1-78278SS
  • Antigen symbol:
  • CK19
  • CK19
  • Antigen name:
  • Cytokeratin 19
  • Cytokeratin 19
  • Antigen synonyms:
  • type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
  • type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 3880
  • 3880
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together