Anti-CXCR4 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-77560B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C Motif) Receptor 4
- Antigen symbol:CXCR4
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:7852
- Antigen synonyms:HM89|LAP-3|WHIM|NPYR|NPYRL|D2S201E|NPY3R|NPYY3R|LCR1|HSY3RR|CD184|WHIMS|LAP3|LESTR|FB22
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human