Order Entry
Spain
ContactUsLinkComponent
Anti-Lamin A + C R453W Mouse Monoclonal Antibody (DyLight® 755) [clone: 12A-2F5]
Anti-Lamin A + C R453W Mouse Monoclonal Antibody (DyLight® 755) [clone: 12A-2F5]
Catalog # NOVUNBP1-77401IR
Supplier:  Novus Biologicals
Anti-Lamin A + C R453W Mouse Monoclonal Antibody (DyLight® 755) [clone: 12A-2F5]
Catalog # NOVUNBP1-77401IR
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lamin A/C
  • Antigen symbol:
    LMNA
  • Clonality:
    Monoclonal
  • Clone:
    12A-2F5
  • Conjugation:
    DyLight® 755
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    4000
  • Antigen synonyms:
    LMNL1|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|LFP|FPLD2|LDP1|CDDC|CMD1A|EMD2|HGPS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: Lamin A + C R453W
Clonality: monoclonal
Clone: 12A-2F5
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human