Order Entry
Spain
ContactUsLinkComponent
Anti-SLC2A4 Rabbit Polyclonal Antibody (Biotin)
Anti-SLC2A4 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-49533B
Supplier:  Novus Biologicals
Anti-SLC2A4 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-49533B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose Transporter 4
  • Antigen symbol:
    GLUT4
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6517
  • Antigen synonyms:
    insulin-responsive glucose transporter type 4|GLUT-4|SLC2A4|glucose transporter type 4|GLUT4|insulin-responsive|facilitated glucose transporter member 4|solute carrier family 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SLC2A4 (solute carrier family 2 (facilitated glucose transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse