Anti-PGAM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
NOVUNBP1-49532H
- Antibody type:Primary
- Antigen name:Phosphoglycerate Mutase 1
- Antigen symbol:PGAM1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5223
- Antigen synonyms:HEL-S-35|PGAMA|PGAM-B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: PGAM1 (phosphoglycerate mutase 1 (brain))
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse